accessibility menu, dialog, popup

UserName

CAS RN 1719-83-1

IUPAC Name: 4,10-dioxatetracyclo[5.5.2.0²,⁶.0⁸,¹²]tetradec-13-ene-3,5,9,11-tetrone
Synonyms: 4,10-dioxatetracyclo[5.5.2.0²,⁶.0⁸,¹²]tetradec-13-ene-3,5,9,11-tetrone
Molecular Weight (g/mol): 248.19
Molecular Formula: C12H8O6
InChi Key: XLOGCGOPKPCECW-UHFFFAOYNA-N
SMILES: O=C1OC(=O)C2C1C1C=CC2C2C1C(=O)OC2=O
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (4)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (4)
  • (1)