accessibility menu, dialog, popup

UserName

CH3OOSOOCH3

CAS RN 17178-10-8

IUPAC Name: 2-methoxyethyl 4-methylbenzene-1-sulfonate
Synonyms: 2-methoxyethyl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 230.28
Molecular Formula: C10H14O4S
InChi Key: TZXJJSAQSRHKCZ-UHFFFAOYSA-N
SMILES: COCCOS(=O)(=O)C1=CC=C(C)C=C1
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results