accessibility menu, dialog, popup

UserName

CAS RN 171058-18-7

IUPAC Name: 1-decyl-3-methyl-1H-imidazol-3-ium chloride
Synonyms: 1-decyl-3-methylimidazolium chloride
Molecular Weight (g/mol): 258.83
Molecular Formula: C14H27ClN2
InChi Key: HTZVLLVRJHAJJF-UHFFFAOYSA-M
SMILES: [Cl-].CCCCCCCCCCN1C=C[N+](C)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results