accessibility menu, dialog, popup

UserName

H3CCH2OOOOCH2OH

CAS RN 1709-71-3

IUPAC Name: 2-hydroxy-3-(prop-2-enoyloxy)propyl 2-methylprop-2-enoate
Synonyms: 2-hydroxy-3-(prop-2-enoyloxy)propyl 2-methylprop-2-enoate
Molecular Weight (g/mol): 214.22
Molecular Formula: C10H14O5
InChi Key: ZODNDDPVCIAZIQ-UHFFFAOYNA-N
SMILES: CC(=C)C(=O)OCC(O)COC(=O)C=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results