accessibility menu, dialog, popup

UserName

CH3ONH2Br

CAS RN 170229-98-8

IUPAC Name: 4-bromo-3-methylbenzamide
Synonyms: 4-bromo-3-methylbenzamide
Molecular Weight (g/mol): 214.06
Molecular Formula: C8H8BrNO
InChi Key: ZPFPOZMNEKPBIF-UHFFFAOYSA-N
SMILES: CC1=C(Br)C=CC(=C1)C(N)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results