accessibility menu, dialog, popup

UserName

CAS RN 1689-64-1

IUPAC Name: 9H-fluoren-9-ol
Synonyms: fluorenol diphenylene carbinol
Molecular Weight (g/mol): 182.22
Molecular Formula: C13H10O
InChi Key: AFMVESZOYKHDBJ-UHFFFAOYSA-N
SMILES: OC1C2=CC=CC=C2C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (3)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)