missing translation for 'onlineSavingsMsg'
Learn More

9-Fluorenol, 98+%

Catalog No. AAL0191714
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0191714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0191714 Supplier Thermo Scientific Chemicals Supplier No. L0191714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1689-64-1 | C13H10O | 182.222 g/mol

9-Fluorenol is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Fluorenol is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Insoluble in water.

Notes
Protect from moisture, heat. Store in cool place. Keep container tightly

Chemical Identifiers

CAS 1689-64-1
Molecular Formula C13H10O
Molecular Weight (g/mol) 182.222
MDL Number MFCD00001135
InChI Key AFMVESZOYKHDBJ-UHFFFAOYSA-N
Synonym 9-hydroxyfluorene, 9-fluorenol, fluoren-9-ol, fluorenol, 9-hydroxy fluorene, diphenylene carbinol, unii-bv0q72r613, hydroxyfluoren, 9-hydroxy-fluoren, 9h-fluorene-9-ol
PubChem CID 74318
ChEBI CHEBI:16904
IUPAC Name 9H-fluoren-9-ol
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)O

Specifications

Melting Point 155°C to 157°C
Quantity 25 g
Beilstein 1869799
Solubility Information Insoluble in water.
Formula Weight 182.22
Percent Purity ≥98%
Chemical Name or Material 9-Fluorenol
EINECSNumber 216-879-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only