accessibility menu, dialog, popup

UserName

CAS RN 1687-53-2

IUPAC Name: 5-amino-2-methoxyphenol
Synonyms: 5-amino-2-methoxyphenol
Molecular Weight (g/mol): 139.15
Molecular Formula: C7H9NO2
InChi Key: BLQFHJKRTDIZLX-UHFFFAOYSA-N
SMILES: COC1=CC=C(N)C=C1O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)