missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-4-methoxyaniline 98.0+%, TCI America™

Catalog No. H06345G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H06345G 5 g
H063425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H06345G Supplier TCI America Supplier No. H06345G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1687-53-2
Molecular Formula C7H9NO2
Molecular Weight (g/mol) 139.154
MDL Number MFCD00010222
InChI Key BLQFHJKRTDIZLX-UHFFFAOYSA-N
Synonym 3-hydroxy-4-methoxyaniline, 5-aminoguaiacol, 2-methoxy-5-aminophenol, phenol, 5-amino-2-methoxy, 5-amino-2-methoxy-phenol, pubchem3112, acmc-209dyt, 3-hydroxy4-methoxyaniline, 5-amino-2-methoxy phenol, 3-hydroxy-4-methoxylaniline
PubChem CID 74314
IUPAC Name 5-amino-2-methoxyphenol
SMILES COC1=C(C=C(C=C1)N)O

Specifications

Melting Point 130°C
Quantity 5 g
Formula Weight 139.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-4-methoxyaniline