accessibility menu, dialog, popup

UserName

CAS RN 16853-74-0

OOOOOZrW

CAS RN 16853-74-0

IUPAC Name: zirconium(4+) tungsten pentaoxidandiide
Synonyms: zirconium tungstate
Molecular Weight (g/mol): 355.06
Molecular Formula: O5WZr
InChi Key: GGUDROIMBGREID-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[O--].[O--].[Zr+4].[W]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results