CAS RN 1672-46-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Digoxigenin, ≥98% (HPLC), MilliporeSigma™ Supelco™
Digoxigenin is a therapeutic drug belonging to the group of cardiac glycosides, widely used in the management of congestive heart failure and other cardiac diseases.