missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Digoxigenin, ≥98% (HPLC), MilliporeSigma™ Supelco™
Description
Featured Industries: Environmental and Food and Beverages
Specifications
Specifications
| CAS | 1672-46-4 |
| Physical Form | Neat |
| Linear Formula | C23H34O5 |
| MDL Number | MFCD00871861 |
| Melting Point | 222°C |
| Synonym | 3 beta,12 beta,14 beta,21-Tetrahydroxy-20(22)-norcholenic acid lactone; 3 beta,12 beta,14-Trihydroxy-5 beta,20(22)-cardenolide; 5 beta,20(22)-Cardenolide-3 beta,12 beta,14-triol; Lanadigigenin |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 390.51 |
| Percent Purity | ≥98% (HPLC) |
| Show More |