accessibility menu, dialog, popup

UserName

CAS RN 1665-00-5

IUPAC Name: dichloro(²H₂)methane
Synonyms: deuterated dichloromethane
Molecular Weight (g/mol): 86.94
Molecular Formula: CH2Cl2
InChi Key: YMWUJEATGCHHMB-DICFDUPASA-N
SMILES: [2H]C([2H])(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results