accessibility menu, dialog, popup

UserName

CAS RN 1658-42-0

H3COON

CAS RN 1658-42-0

IUPAC Name: methyl 2-(pyridin-2-yl)acetate
Synonyms: methyl 2-(pyridin-2-yl)acetate
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: ORAKNQSHWMHCEY-UHFFFAOYSA-N
SMILES: COC(=O)CC1=CC=CC=N1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results