accessibility menu, dialog, popup

UserName

FFFFFOOOOFFFFF

CAS RN 16536-48-4

IUPAC Name: bis(2,3,4,5,6-pentafluorophenyl) oxalate
Synonyms: bis(2,3,4,5,6-pentafluorophenyl) oxalate
Molecular Weight (g/mol): 422.13
Molecular Formula: C14F10O4
InChi Key: YMTUYHWOWDFXOX-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(OC(=O)C(=O)OC2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results