missing translation for 'onlineSavingsMsg'
Learn More

Bis(pentafluorophenyl) Oxalate [Chemiluminescence Reagent], TCI America™

Catalog No. A53051G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A53051G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A53051G Supplier TCI America Supplier No. A53051G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16536-48-4
Molecular Formula C14F10O4
Molecular Weight (g/mol) 422.13
MDL Number MFCD00143774
InChI Key YMTUYHWOWDFXOX-UHFFFAOYSA-N
Synonym PFPO
PubChem CID 2736057
IUPAC Name bis(2,3,4,5,6-pentafluorophenyl) oxalate
SMILES FC1=C(F)C(F)=C(OC(=O)C(=O)OC2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F

Specifications

Color White
Quantity 1 g
Formula Weight 422.13
Physical Form Crystalline Powder
Chemical Name or Material Bis(pentafluorophenyl) Oxalate [Chemiluminescence Reagent]