accessibility menu, dialog, popup

UserName

CAS RN 16534-12-6

IUPAC Name: 4-bromo-3,5-dihydroxybenzoic acid
Synonyms: 4-bromo-3,5-dihydroxybenzoic acid
Molecular Weight (g/mol): 233.02
Molecular Formula: C7H5BrO4
InChi Key: NUTRHYYFCDEALP-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(O)=C(Br)C(O)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)