accessibility menu, dialog, popup

UserName

PCl

CAS RN 16523-54-9

IUPAC Name: chlorodicyclohexylphosphane
Synonyms: chlorodicyclohexylphosphane
Molecular Weight (g/mol): 232.73
Molecular Formula: C12H22ClP
InChi Key: AKJFBIZAEPTXIL-UHFFFAOYSA-N
SMILES: ClP(C1CCCCC1)C1CCCCC1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)