accessibility menu, dialog, popup

UserName

CAS RN 164470-64-8

IUPAC Name: 4-[({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)methyl]benzoic acid
Synonyms: 4-({[(9H-fluoren-9-ylmethoxy)carbonyl]amino}methyl)benzoic acid
Molecular Weight (g/mol): 373.41
Molecular Formula: C23H19NO4
InChi Key: JRHUROPSUJVMNH-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(CNC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)C=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)