accessibility menu, dialog, popup

UserName

CH3NH3CFNH3CH3C

CAS RN 164298-23-1

IUPAC Name: [(dimethylamino)(fluoro)methylidene]dimethylazanium
Synonyms: [(dimethylamino)(fluoro)methylidene]dimethylazanium
Molecular Weight (g/mol): 119.16
Molecular Formula: C5H12FN2
InChi Key: JILUGXGJYPRQPA-UHFFFAOYSA-N
SMILES: CN(C)C(F)=[N+](C)C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)