accessibility menu, dialog, popup

UserName

CAS RN 16357-59-8

IUPAC Name: ethyl 2-ethoxy-1,2-dihydroquinoline-1-carboxylate
Synonyms: ethyl 2-ethoxy-2H-quinoline-1-carboxylate
Molecular Weight (g/mol): 247.29
Molecular Formula: C14H17NO3
InChi Key: GKQLYSROISKDLL-UHFFFAOYNA-N
SMILES: CCOC1C=CC2=CC=CC=C2N1C(=O)OCC
Percent Purity 
  • (1)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (6)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (4)