accessibility menu, dialog, popup

UserName

CAS RN 163521-08-2

NNNHNOOH2NHCl

CAS RN 163521-08-2

IUPAC Name: 5-{4-[4-(5-cyano-1H-indol-3-yl)butyl]piperazin-1-yl}-1-benzofuran-2-carboxamide hydrochloride
Synonyms: vilazodone hydrochloride
Molecular Weight (g/mol): 477.99
Molecular Formula: C26H28ClN5O2
InChi Key: RPZBRGFNBNQSOP-UHFFFAOYSA-N
SMILES: Cl.NC(=O)C1=CC2=CC(=CC=C2O1)N1CCN(CCCCC2=CNC3=CC=C(C=C23)C#N)CC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results