accessibility menu, dialog, popup

UserName

CAS RN 1635-61-6

IUPAC Name: 5-chloro-2-nitroaniline
Synonyms: 5-chloro-2-nitroaniline
Molecular Weight (g/mol): 172.57
Molecular Formula: C6H5ClN2O2
InChi Key: ZCWXYZBQDNFULS-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=CC=C1[N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)