accessibility menu, dialog, popup

UserName

CAS RN 16348-49-5

IUPAC Name: 2-amino-N'-hydroxybenzene-1-carboximidamide
Synonyms: 2-amino-N'-hydroxybenzenecarboximidamide
Molecular Weight (g/mol): 151.17
Molecular Formula: C7H9N3O
InChi Key: CFZHYRNQLHEHJS-UHFFFAOYSA-N
SMILES: NC(=NO)C1=CC=CC=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results