accessibility menu, dialog, popup

UserName

CH3OSOOOOHK

CAS RN 16241-25-1

IUPAC Name: potassium 4-hydroxy-3-methoxybenzene-1-sulfonate
Synonyms: potassium 4-hydroxy-3-methoxybenzenesulfonate
Molecular Weight (g/mol): 242.29
Molecular Formula: C7H7KO5S
InChi Key: QFRKWSPTCBGLSU-UHFFFAOYSA-M
SMILES: [K+].COC1=C(O)C=CC(=C1)S([O-])(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results