accessibility menu, dialog, popup

UserName

FFOOOHNClNCl

CAS RN 162401-32-3

IUPAC Name: 3-(cyclopropylmethoxy)-N-(3,5-dichloropyridin-4-yl)-4-(difluoromethoxy)benzamide
Synonyms: roflumilast
Molecular Weight (g/mol): 403.21
Molecular Formula: C17H14Cl2F2N2O3
InChi Key: MNDBXUUTURYVHR-UHFFFAOYSA-N
SMILES: FC(F)OC1=CC=C(C=C1OCC1CC1)C(=O)NC1=C(Cl)C=NC=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results