accessibility menu, dialog, popup

UserName

CAS RN 162359-56-0

IUPAC Name: hydrogen 2-amino-2-[2-(4-octylphenyl)ethyl]propane-1,3-diol chloride
Synonyms: hydrogen fingolimod chloride
Molecular Weight (g/mol): 343.94
Molecular Formula: C19H34ClNO2
InChi Key: SWZTYAVBMYWFGS-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCCCCCCCC1=CC=C(CCC(N)(CO)CO)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Product Type 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results