missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fingolimod Hydrochloride 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 110°C |
| Color | White |
| Quantity | 200 mg |
| Formula Weight | 343.94 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Fingolimod Hydrochloride |
Safety and Handling
| RTECSNumber | TY3130000 |
| TSCA | No |
| Recommended Storage | Freezer (-20 °C) |