accessibility menu, dialog, popup

UserName

CAS RN 1623-92-3

IUPAC Name: 4-phenoxybenzene-1-sulfonyl chloride
Synonyms: 4-phenoxybenzenesulfonyl chloride
Molecular Weight (g/mol): 268.71
Molecular Formula: C12H9ClO3S
InChi Key: QIZPONOMFWAPRR-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=C(OC2=CC=CC=C2)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results