CAS RN 16208-17-6
IUPAC Name:
2,2-dihydroxy-1-(4-methoxyphenyl)ethan-1-one
Synonyms:
2,2-dihydroxy-1-(4-methoxyphenyl)ethanone
Molecular Weight (g/mol):
182.18
Molecular Formula:
C9H10O4
InChi Key:
NLUFPYPVEHQAQN-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C=C1)C(=O)C(O)O