accessibility menu, dialog, popup

UserName

CAS RN 1619-57-4

IUPAC Name: ethyl 2,2-diethyl-3-oxobutanoate
Synonyms: ethyl 2,2-diethyl-3-oxobutanoate
Molecular Weight (g/mol): 186.25
Molecular Formula: C10H18O3
InChi Key: WEIQRLLXVVSKIL-UHFFFAOYSA-N
SMILES: CCOC(=O)C(CC)(CC)C(C)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results