accessibility menu, dialog, popup

UserName

FOFF

CAS RN 161793-17-5

IUPAC Name: 2,3,4-trifluorobenzaldehyde
Synonyms: 2,3,4-trifluorobenzaldehyde
Molecular Weight (g/mol): 160.1
Molecular Formula: C7H3F3O
InChi Key: UQEDGFZRPSAHLC-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=O)C(F)=C1F
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)