accessibility menu, dialog, popup

UserName

CAS RN 161091-49-2

CH3NH3CNH2Br

CAS RN 161091-49-2

IUPAC Name: 3-bromo-5,6-dimethylpyridin-2-amine
Synonyms: 3-bromo-5,6-dimethylpyridin-2-amine
Molecular Weight (g/mol): 201.07
Molecular Formula: C7H9BrN2
InChi Key: PZIWHEZMEJMJLP-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=C(N)N=C1C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results