accessibility menu, dialog, popup

UserName

CAS RN 1609-93-4

IUPAC Name: (2Z)-3-chloroprop-2-enoic acid
Synonyms: cis-3-chloroacrylic acid C3H3ClO2
Molecular Weight (g/mol): 106.51
Molecular Formula: C3H3ClO2
InChi Key: MHMUCYJKZUZMNJ-UPHRSURJSA-N
SMILES: OC(=O)\C=C/Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)