accessibility menu, dialog, popup

UserName

NH2

CAS RN 1606-67-3

IUPAC Name: pyren-1-amine
Synonyms: 1-pyrenamine
Molecular Weight (g/mol): 217.27
Molecular Formula: C16H11N
InChi Key: YZVWKHVRBDQPMQ-UHFFFAOYSA-N
SMILES: NC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34
Percent Purity 
  • (3)
  • (4)
Physical Form 
  • (3)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (4)
  • (3)
  • (4)
  • (3)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)