accessibility menu, dialog, popup

UserName

NN

CAS RN 15937-81-2

IUPAC Name: 2-[3-(pyridin-2-yl)propyl]pyridine
Synonyms: 2-[3-(pyridin-2-yl)propyl]pyridine
Molecular Weight (g/mol): 198.27
Molecular Formula: C13H14N2
InChi Key: UHPWYBATPHCMGD-UHFFFAOYSA-N
SMILES: C(CC1=CC=CC=N1)CC1=CC=CC=N1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results