CAS RN 15931-15-4
IUPAC Name:
3-fluoro-2-methylpyridine
Synonyms:
3-fluoro-2-methylpyridine
Molecular Weight (g/mol):
111.12
Molecular Formula:
C6H6FN
InChi Key:
IRIHSZDBTACXCT-UHFFFAOYSA-N
SMILES:
CC1=C(F)C=CC=N1
1
–
1
of
1
results
1
Promotions Available
3-Fluoro-2-methylpyridine, 98%
3-Fluoro-2-methylpyridine, 98%
CAS: 15931-15-4 Molecular Formula: C6H6FN Molecular Weight (g/mol): 111.119 MDL Number: MFCD08705710 InChI Key: IRIHSZDBTACXCT-UHFFFAOYSA-N Synonym: 3-fluoro-2-picoline,pyridine, 3-fluoro-2-methyl,2-methyl-3-fluoropyridine,3-fluoro-2-methyl pyridine,3-fluoro-2-methyl-pyridine,fluoropicoline,pubchem14381,acmc-209xci,ksc530a3b,pyridine,3-fluoro-2-methyl PubChem CID: 21613172 IUPAC Name: 3-fluoro-2-methylpyridine SMILES: CC1=C(C=CC=N1)F