accessibility menu, dialog, popup

UserName

CAS RN 1591-30-6

IUPAC Name: [1,1'-biphenyl]-4,4'-dicarbonitrile
Synonyms: [1,1'-biphenyl]-4,4'-dicarbonitrile
Molecular Weight (g/mol): 204.23
Molecular Formula: C14H8N2
InChi Key: KAXYYLCSSXFXKR-UHFFFAOYSA-N
SMILES: N#CC1=CC=C(C=C1)C1=CC=C(C=C1)C#N
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)