missing translation for 'onlineSavingsMsg'
Learn More

Biphenyl-4,4'-dicarbonitrile, 98%

Catalog No. AAL0345006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0345006 5 g
AAL0345003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0345006 Supplier Thermo Scientific Chemicals Supplier No. L0345006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1591-30-6 | C14H8N2 | 204.232 g/mol

Biphenyl-4,4'-dicarbonitrile is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biphenyl-4,4′-dicarbonitrile is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 1591-30-6
Molecular Formula C14H8N2
Molecular Weight (g/mol) 204.232
MDL Number MFCD00013805
InChI Key KAXYYLCSSXFXKR-UHFFFAOYSA-N
Synonym 4,4'-biphenyldicarbonitrile, 1,1'-biphenyl-4,4'-dicarbonitrile, 4,4'-dicyanobiphenyl, biphenyl-4,4'-dicarbonitrile, 4,4'-dicyanodiphenyl, 4,4'-diphenyldicarbonitrile, 4-4-cyanophenyl benzonitrile, 4-4-cyanophenyl benzenecarbonitrile
PubChem CID 15321
IUPAC Name 4-(4-cyanophenyl)benzonitrile
SMILES C1=CC(=CC=C1C#N)C2=CC=C(C=C2)C#N

Specifications

Melting Point 232°C to 236°C
Quantity 5 g
UN Number UN3439
Beilstein 1869917
Solubility Information Insoluble in water.
Formula Weight 204.23
Percent Purity 98%
Chemical Name or Material Biphenyl-4,4'-dicarbonitrile
Hazard Category H302+H332
Hazard Statement GHS H Statement
H331-H302
Toxic if inhaled.
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 216-468-6
RTECSNumber DV2625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only