missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Biphenyldicarbonitrile 97.0+%, TCI America™

Catalog No. B11811G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B11811G Supplier TCI America Supplier No. B11811G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1591-30-6
Molecular Formula C14H8N2
Molecular Weight (g/mol) 204.232
MDL Number MFCD00013805
InChI Key KAXYYLCSSXFXKR-UHFFFAOYSA-N
Synonym 4,4'-biphenyldicarbonitrile, 1,1'-biphenyl-4,4'-dicarbonitrile, 4,4'-dicyanobiphenyl, biphenyl-4,4'-dicarbonitrile, 4,4'-dicyanodiphenyl, 4,4'-diphenyldicarbonitrile, 4-4-cyanophenyl benzonitrile, 4-4-cyanophenyl benzenecarbonitrile
PubChem CID 15321
IUPAC Name 4-(4-cyanophenyl)benzonitrile
SMILES C1=CC(=CC=C1C#N)C2=CC=C(C=C2)C#N

Specifications

Melting Point 238°C
Color Yellow
Quantity 1 g
Formula Weight 204.23
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Biphenyldicarbonitrile
RTECSNumber DV2625000
TSCA Yes