accessibility menu, dialog, popup

UserName

H3CONOI

CAS RN 158669-26-2

IUPAC Name: 4-iodo-2-methoxypyridine-3-carbaldehyde
Synonyms: 4-iodo-2-methoxypyridine-3-carbaldehyde
Molecular Weight (g/mol): 263.03
Molecular Formula: C7H6INO2
InChi Key: GNBKAOHTTIVAMT-UHFFFAOYSA-N
SMILES: COC1=NC=CC(I)=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results