accessibility menu, dialog, popup

UserName

CH3OCH3

CAS RN 1585-06-4

IUPAC Name: 1-ethoxy-4-ethylbenzene
Synonyms: P-ethylphenetole
Molecular Weight (g/mol): 150.22
Molecular Formula: C10H14O
InChi Key: BQBROHBMIBOPFU-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(CC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results