accessibility menu, dialog, popup

UserName

CAS RN 15823-43-5

IUPAC Name: hafnium sulfate
Synonyms: hafnium sulfate
Molecular Weight (g/mol): 274.55
Molecular Formula: HfO4S
InChi Key: KOELDGKLEFTHGN-UHFFFAOYSA-L
SMILES: [Hf].[O-]S([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results