accessibility menu, dialog, popup

UserName

CAS RN 1576-87-0

IUPAC Name: (2E)-pent-2-enal
Synonyms: 2-pentenal
Molecular Weight (g/mol): 84.12
Molecular Formula: C5H8O
InChi Key: DTCCTIQRPGSLPT-ONEGZZNKSA-N
SMILES: CC\C=C\C=O
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (4)