accessibility menu, dialog, popup

UserName

CAS RN 1571-72-8

IUPAC Name: 3-amino-4-hydroxybenzoic acid
Synonyms: cheetan
Molecular Weight (g/mol): 153.14
Molecular Formula: C7H7NO3
InChi Key: MRBKRZAPGUCWOS-UHFFFAOYSA-N
SMILES: NC1=CC(=CC=C1O)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)