accessibility menu, dialog, popup

UserName

H2OClCH3N(R)S(R)HNO(R)H2NOHOOH

CAS RN 15686-71-2

IUPAC Name: hydrogen (6R,7R)-7-[(2R)-2-amino-2-phenylacetamido]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid hydrate chloride
Synonyms: cefalexin
Molecular Weight (g/mol): 401.86
Molecular Formula: C16H20ClN3O5S
InChi Key: YHJDZIQOCSDIQU-OEDJVVDHSA-N
SMILES: [H+].O.[Cl-].CC1=C(N2[C@H](SC1)[C@H](NC(=O)[C@H](N)C1=CC=CC=C1)C2=O)C(O)=O
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Type 
  • (1)
  • (2)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)