missing translation for 'onlineSavingsMsg'
Learn More

Cephalexin Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013255
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013255 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013255 Supplier Merck Emd Millipore Supplier No. PHR18481G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Cephalexin is a cephalosporine which is categorized under the class of β-lactam antibiotics. It is utilized for regulating the effects of Gram-positive and Gram-negative bacteria. It favors the lysis of bacterial cell wall by inhibiting the synthesis of essential structural components. Cephalexin belongs to the safest group of broad-spectrum bactericidal antimicrobial agents.

Specifications

CAS 15686-71-2
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C16H17N3O4 S
MDL Number MFCD00056877
Synonym Cefalexin; 7-(D-alpha-amino-phenylacetamido)-3-methyl-3-cepheme-4-carboxylic acid
Type Certified Reference Material
Quantity 1 g
Formula Weight 347.39
Grade Certified Reference Material