accessibility menu, dialog, popup

UserName

CAS RN 15681-89-7

IUPAC Name: sodium boranuide
Synonyms: sodium borohydride|NaBH4|sodium tetrahydroborate|borohydrure de sodium|sodium tetrahydroborate(1-)|sodium tetrahydridoborate(1-)
Molecular Weight (g/mol): 37.83
Molecular Formula: BH4Na
InChi Key: YOQDYZUWIQVZSF-UHFFFAOYSA-N
SMILES: [BH4-].[Na+]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results