accessibility menu, dialog, popup

UserName

CAS RN 156682-52-9

BrFFBr

CAS RN 156682-52-9

IUPAC Name: 1,4-dibromo-2,3-difluorobenzene
Synonyms: 1,4-dibromo-2,3-difluorobenzene
Molecular Weight (g/mol): 271.89
Molecular Formula: C6H2Br2F2
InChi Key: RGXGEFSBDPGCEU-UHFFFAOYSA-N
SMILES: FC1=C(Br)C=CC(Br)=C1F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results