accessibility menu, dialog, popup

UserName

CH3OBOHOH

CAS RN 156641-98-4

IUPAC Name: (6-methoxynaphthalen-2-yl)boronic acid
Synonyms: 6-methoxynaphthalen-2-ylboronic acid
Molecular Weight (g/mol): 202.02
Molecular Formula: C11H11BO3
InChi Key: GZFAVYWCPSMLCM-UHFFFAOYSA-N
SMILES: COC1=CC2=CC=C(C=C2C=C1)B(O)O
Quantity 
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)